Merck
  • Conditional steroidogenic cell-targeted deletion of TSPO unveils a crucial role in viability and hormone-dependent steroid formation.
Conditional steroidogenic cell-targeted deletion of TSPO unveils a crucial role in viability and hormone-dependent steroid formation.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-04)
Jinjiang Fan, Enrico Campioli, Andrew Midzak, Martine Culty, Vassilios Papadopoulos
ABSTRACT
Translocator protein (TSPO) is a key member of the mitochondrial cholesterol transport complex in steroidogenic tissues. To assess the function of TSPO, we generated two lines of Cre-mediated Tspo conditional knockout (cKO) mice. First, gonadal somatic cell-targeting Amhr2-Cre mice were crossed with Tspo-floxed mice to obtain F1 Tspo Amhr2 cKO mice (Tspo(fl/fl);Amhr2-Cre(/+)). The unexpected Mendelian ratio of 4.4% cKO mice was confirmed by genotyping of 12.5-day-postcoitum (dpc) embryos. As Amhr2-Cre is expressed in gonads at 12.5 dpc, these findings suggest preimplantation selection of embryos. Analysis of expression databases revealed elevated levels of Amhr2 in two- and eight-cell zygotes, suggesting ectopic Tspo silencing before the morula stage and demonstrating elevated embryonic lethality and involvement of TSPO in embryonic development. To circumvent this issue, steroidogenic cell-targeting Nr5a1-Cre mice were crossed with Tspo-floxed mice. The resulting Tspo(fl/fl);Nr5a1-Cre(/+) mice were born at a normal Mendelian ratio. Nr5a1-driven Tspo cKO mice exhibited highly reduced Tspo levels in adrenal cortex and gonads. Treatment of mice with human chorionic gonadotropin (hCG) resulted in increased circulating testosterone levels despite extensive lipid droplet depletion. In contrast, Nr5a1-driven Tspo cKO mice lost their ability to form corticosterone in response to adrenocorticotropic hormone (ACTH). Important for ACTH-dependent steroidogenesis, Mc2r, Stard1, and Cypa11a1 levels were unaffected, whereas Scarb1 levels were increased and accumulation of lipid droplets was observed, indicative of a blockade of cholesterol utilization for steroidogenesis. TSPO expression in the adrenal medulla and increased epinephrine production were also observed. In conclusion, TSPO was found necessary for preimplantation embryo development and ACTH-stimulated steroid biosynthesis.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications