Skip to Content
Merck
All Photos(3)

Key Documents

357456

Sigma-Aldrich

Copper

powder (dendritic), <45 μm, 99.7% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$178.00
500 G
$338.00

$178.00


Estimated to ship onMarch 17, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$178.00
500 G
$338.00

About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
EC Number:
MDL number:
UNSPSC Code:
12141711
PubChem Substance ID:
NACRES:
NA.23

$178.00


Estimated to ship onMarch 17, 2025


Request a Bulk Order

Quality Level

Assay

99.7% trace metals basis

form

powder (dendritic)

resistivity

1.673 μΩ-cm, 20°C

particle size

<45 μm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Related Categories

General description

Copper powder (dendritic) is also known as an electrolytic copper powder (ECP) which has a dendritic shape with a particle size that is less than 4.5 μm. The dendritic shape of the copper particles can be produced by electrolytic deposition method.[1][2]

Application

ECP can be used as a catalyst in the production of biodiesel by transesterification of soybean oil and partial hydrogenation of polyunsaturated compounds.[3] It can also be used to form a metal composite with carbon nanotubes for the formation of high strength copper alloys.[1]

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formation of dendritic copper deposit in industrial electrolysis
Osipova ML, et al.
Powder Metallurgy and Metal Ceramics, 49(5-6), 253-259 (2010)
Effect of size and shape of metal particles to improve hardness and electrical properties of carbon nanotube reinforced copper and copper alloy composites
Uddin SM, et al.
Composites Science and Technology, 70(16), 2253-2257 (2010)
Biodiesel production from highly unsaturated feedstock via simultaneous transesterification and partial hydrogenation in supercritical methanol
Shin H, et al.
Journal of Supercritical Fluids, 82(5-6), 251-255 (2013)
Hiroshi Sato et al.
Science (New York, N.Y.), 343(6167), 167-170 (2013-12-18)
Carbon monoxide (CO) produced in many large-scale industrial oxidation processes is difficult to separate from nitrogen (N2), and afterward, CO is further oxidized to carbon dioxide. Here, we report a soft nanoporous crystalline material that selectively adsorbs CO with adaptable
Seonah Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 149-154 (2013-12-18)
Lytic polysaccharide monooxygenases (LPMOs) exhibit a mononuclear copper-containing active site and use dioxygen and a reducing agent to oxidatively cleave glycosidic linkages in polysaccharides. LPMOs represent a unique paradigm in carbohydrate turnover and exhibit synergy with hydrolytic enzymes in biomass

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service