Skip to Content
Merck
All Photos(1)

Documents

348481

Sigma-Aldrich

Potassium nitrate-15N

10 atom % 15N

Synonym(s):

15N Labeled KNO3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K15NO3
CAS Number:
Molecular Weight:
102.10
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

10 atom % 15N

form

solid

mol wt

101.2 by atom % calculation (10 atom % 15N)

mp

334 °C (lit.)

density

2.132 g/mL

application(s)

agriculture

mass shift

M+1

SMILES string

[K+].[O-][15N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1/i;1+1

InChI key

FGIUAXJPYTZDNR-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service