Skip to Content
Merck
All Photos(1)

Key Documents

347779

Sigma-Aldrich

Isoamyltriphenylphosphonium bromide

98%

Synonym(s):

(3-Methylbutyl)triphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$91.60

$91.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$91.60

About This Item

Linear Formula:
(CH3)2CHCH2CH2P(C6H5)3Br
CAS Number:
Molecular Weight:
413.33
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$91.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

157-159 °C (lit.)

functional group

phosphine

SMILES string

[Br-].CC(C)CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H26P.BrH/c1-20(2)18-19-24(21-12-6-3-7-13-21,22-14-8-4-9-15-22)23-16-10-5-11-17-23;/h3-17,20H,18-19H2,1-2H3;1H/q+1;/p-1

InChI key

GZLGTVRDLCJQTO-UHFFFAOYSA-M

Application

Reactant for:
  • Preparation of 9,11-secosterol analogs as antitumor agents
  • Synthesis of antiepileptic, ceramide I, and its triacetyl derivatives via olefin cross metathesis
  • Preparation of meso-epoxides via substitution of alkyl bromides with triphenylphosphine followed by Wittig olefination and epoxidation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service