Skip to Content
Merck
All Photos(1)

Key Documents

347590

Sigma-Aldrich

2-[4-(Dimethylamino)phenyl]ethanol

≥99%

Synonym(s):

4-(Dimethylamino)phenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CH2CH2OH
CAS Number:
Molecular Weight:
165.23
Beilstein:
2717032
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
≥99%

Assay

≥99%

form

solid

mp

54-58 °C (lit.)

SMILES string

CN(C)c1ccc(CCO)cc1

InChI

1S/C10H15NO/c1-11(2)10-5-3-9(4-6-10)7-8-12/h3-6,12H,7-8H2,1-2H3

InChI key

CDTPAAZQBPSVGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-[4-(Dimethylamino)phenyl]ethanol (4-(Dimethylamino)phenethyl alcohol) is a more effective accelerator than N,N-dimethyl-p-toluidine (TD) for bone cement curing.[1]

Application

2-[4-(Dimethylamino)phenyl]ethanol (4-(Dimethylamino)phenethyl alcohol) may be used as accelerator to investigate the the curing of bone cement at molecular level by electron spin resonance (ESR) spectroscopy.[1] 2-[4-(Dimethylamino)phenyl]ethanol (4-(N,N-dimethylamino)phenethyl alcohol) may be used to compare the efficiency of different camphorquinone (CQ)/amine photo-initiating systems for the photopolymerization of a model dental resin.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F F Oldfield et al.
Journal of biomedical materials research, 44(4), 436-445 (1999-07-09)
Electron spin resonance (ESR) spectroscopy was used to gain insight at the molecular level into the curing of bone cement. Methyl methacrylate was polymerized using a N,N-dimethyl-p-toluidine (TD)/benzoyl peroxide (BPO) redox system in the presence of polymethyl methacrylate (PMMA) powder.
Walter F Schroeder et al.
Dental materials : official publication of the Academy of Dental Materials, 23(10), 1313-1321 (2006-12-19)
The aim of this study was to compare the efficiency of different camphorquinone (CQ)/amine photo-initiating systems for the photopolymerization of a model dental resin based in Bis-GMA/TEGDMA. The monomer conversion versus exposure time was measured in resins containing different types

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service