Skip to Content
Merck
All Photos(1)

Key Documents

324582

Sigma-Aldrich

Iodoethane-d5

99.5 atom % D, contains copper as stabilizer

Synonym(s):

Ethyl-d5 iodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$299.00
5 G
$991.00

$299.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$299.00
5 G
$991.00

About This Item

Linear Formula:
CD3CD2I
CAS Number:
Molecular Weight:
161.00
Beilstein:
1840101
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

$299.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99.5 atom % D

Quality Level

Assay

99% (CP)

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.5091 (lit.)

bp

69-73 °C (lit.)

mp

-108 °C (lit.)

density

2.013 g/mL at 25 °C

application(s)

pharmaceutical

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])I

InChI

1S/C2H5I/c1-2-3/h2H2,1H3/i1D3,2D2

InChI key

HVTICUPFWKNHNG-ZBJDZAJPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup - (External MSDS)(in analogy to similar products)

Flash Point(C)

53 °C - closed cup - (External MSDS)(in analogy to similar products)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of Chromatography A, 688, 243-243 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service