Skip to Content
Merck
All Photos(1)

Key Documents

346292

Sigma-Aldrich

4,4-Dimethyl-2-phenyl-2-oxazoline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5322 (lit.)

bp

124 °C/20 mmHg (lit.)

mp

20-24 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

SMILES string

CC1(C)COC(=N1)c2ccccc2

InChI

1S/C11H13NO/c1-11(2)8-13-10(12-11)9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

UGNSMKDDFAUGFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4-Dimethyl-2-phenyl-2-oxazoline is an 2-aryl-2-oxazoline and has been tested as as ligand for the Suzuki coupling reaction of aryl bromides and arylboronic acids.[1] 4,4-Dimethyl-2-phenyl-2-oxazoline forms mildly hygroscopic complexes, of general formulae [ZnX2(ox)2] (ox =4,4-Dimethyl-2-phenyl-2-oxazoline), with ether solutions of [ZnX2] (X = Cl, Br, I).Except ZnI2, which does not form an isolable complex due to steric reasons.[2]

Application

4,4-Dimethyl-2-phenyl-2-oxazoline may be used in the synthesis of an ester-ammonium salt via rupture of oxazoline ring.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxazoles XXI. Three Ester Ammonium Salts Resulting from the Rupture of an Oxazoline Ring.
Gossage RA, et al.
Journal of Chemical Crystallography, 40(3), 272-277 (2010)
Oxazoline chemistry-Part IV: Synthesis and characterization of oxazoline complexes of the zinc halides.
Barclay TM, et al.
Canadian Journal of Chemistry, 81(12), 1482-1491 (2003)
Pd (OAc)2/2-aryl-2-oxazolines catalyzed Suzuki coupling reactions of aryl bromides and arylboronic acids.
Tao B and Boykin DW.
Tetrahedron Letters, 43(28), 4955-4957 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service