Skip to Content
Merck
All Photos(3)

Key Documents

345849

Sigma-Aldrich

Dibenzoyl-L-tartaric acid

98%

Synonym(s):

L-DBTA, (−)-O,O′-Dibenzoyl-L-tartaric acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$78.50
100 G
$140.00

$78.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$78.50
100 G
$140.00

About This Item

Linear Formula:
[C6H5CO2CH(CO2H)-]2
CAS Number:
Molecular Weight:
358.30
Beilstein:
709854
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$78.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]20/D −116°, c = 9 in ethanol

mp

152-155 °C (lit.)

functional group

carboxylic acid
ester
phenyl

SMILES string

OC(=O)[C@H](OC(=O)c1ccccc1)[C@@H](OC(=O)c2ccccc2)C(O)=O

InChI

1S/C18H14O8/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12/h1-10,13-14H,(H,19,20)(H,21,22)/t13-,14-/m1/s1

InChI key

YONLFQNRGZXBBF-ZIAGYGMSSA-N

Application

Dibenzoyl-L-tartaric acid may be used as a chiral resolving agent for the resolution of racemic Troger base.[1] It may also be used as a ligand to synthesize chiral transition metal complexes, which have potential utility in organic asymmetric catalysis.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient method for the synthesis and resolution of Troger base.
Satishkumar S and Periasamy M.
Tetrahedron Asymmetry, 17(7), 1116-1119 (2006)
Transition metal complexes of dibenzoyl-L-tartaric acid (db-L-tarH2) and L-tartaric acid (L-tarH2); X-ray crystal structure of {[Cu(L-tar)(phen)]?6H2O}n (phen=1,10-phenanthroline).
McCann M, et al.
Polyhedron, 16(20), 3655-3661 (1997)
C Q Cao et al.
European journal of pharmacology, 418(1-2), 79-87 (2001-05-04)
Due to low central nervous system (CNS) bioavailability of delta-opioid peptides, little is known about the effect of systemic administration of delta-opioid receptor ligands. The present study examined the effect of non-peptidergic delta-opioid receptor agonists, (+)-4-[(alphaR)-alpha-((2R,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide (SNC80) and (-)dibenzoyl-L-tartaric acid
Emily M Jutkiewicz et al.
The Journal of pharmacology and experimental therapeutics, 309(1), 173-181 (2004-01-15)
The diarylpiperazine delta-opioid agonist SNC80 [(+)-4-[(alphaR)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-methoxyphenyl)methyl]-N,N-diethylbenzamide] produces convulsions, antidepressant-like effects, and locomotor stimulation in rats. The present study compared the behavioral effects in Sprague-Dawley rats of SNC80 with its two derivatives, SNC86 [(+)-4-[alpha(R)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-hydroxyphenyl)methyl]-N,N-diethylbenzamide] and SNC162 [(+)-4-[(alphaR)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-phenyl)methyl]-N,N-diethylbenzamide], which differ by one
Stanley I Goldberg
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 43(1), 31-37 (2013-01-25)
Construction and operation of a laboratory model, which combines the lately discovered enantioenrichment method of the author (2007) with the sun-powered evaporative pumping process of Hsu and Siegenthaler (Sedimentology 12:11-25 1969), is described. The model operated continuously for 120 days

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service