Skip to Content
Merck
All Photos(2)

Key Documents

302813

Sigma-Aldrich

(+)-O,O′-Di-p-toluoyl-D-tartaric acid

made from synthetic tartaric acid, 97%

Synonym(s):

Di-p-toluoyl-D-tartaric acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$212.00
100 G
$547.00

$212.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$212.00
100 G
$547.00

About This Item

Linear Formula:
[CH3C6H4CO2CH(CO2H)-]2
CAS Number:
Molecular Weight:
386.35
Beilstein:
3225584
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$212.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

optical activity

[α]19/D +138°, c = 1 in ethanol

mp

169-171 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

Cc1ccc(cc1)C(=O)O[C@@H]([C@H](OC(=O)c2ccc(C)cc2)C(O)=O)C(O)=O

InChI

1S/C20H18O8/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/t15-,16-/m0/s1

InChI key

CMIBUZBMZCBCAT-HOTGVXAUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service