Skip to Content
Merck
All Photos(2)

Key Documents

344575

Sigma-Aldrich

4-Hydroxy-3-nitrobenzonitrile

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$447.00

$447.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$447.00

About This Item

Linear Formula:
HOC6H3(NO2)CN
CAS Number:
Molecular Weight:
164.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$447.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

146-148 °C (lit.)

functional group

nitrile
nitro

SMILES string

Oc1ccc(cc1[N+]([O-])=O)C#N

InChI

1S/C7H4N2O3/c8-4-5-1-2-7(10)6(3-5)9(11)12/h1-3,10H

InChI key

INBLGVOPOSGVTA-UHFFFAOYSA-N

General description

4-Hydroxy-3-nitrobenzonitrile is one of the main photoproduct formed during hotolytic destruction of herbicide bromoxynil.[1]

Application

4-Hydroxy-3-nitrobenzonitrile may be used in the synthesis of:
  • 3-azido-4-methoxybenzonitrile[2]
  • 4-methoxy-3-nitrobenzonitrile[2]
  • ethyl [2,2-dimethyl-6-(Δ2-thiazolin-2-yl)-4H-l,4-benzoxazin-3-one-4-yl]butyrate[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemistry of halogenated benzene derivatives. part xii.* effects of sodium nitrite on the environmental phototransformation of bromoxynil (3, 5-dibromo-4-hydroxybenzonitrile) herbicide in water: The photoincorporation of nitrite ions into bromoxynil.
Kochany J and Choudhry GG.
Toxicological & Environmental Chemistry, 27(4), 225-239 (1990)
X-Ray Structural Analysis of Ethyl [2, 2-dimethyl-6-(?2-thiazolin-2-yl)-4H-1, 4-benzoxazin-3-one-4-yl] butyrate.
Caliendo G, et al.
Structural Chemistry, 9(2), 121-127 (1998)
Stanislav A Bakunov et al.
Journal of medicinal chemistry, 53(1), 254-272 (2009-11-26)
Novel dicationic triazoles 1-60 were synthesized by the Pinner method from the corresponding dinitriles, prepared via the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC). The type and the placement of cationic moieties as well as the nature of aromatic substituents influenced in vitro

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service