Skip to Content
Merck
All Photos(1)

Key Documents

342548

Sigma-Aldrich

3-(Trifluoromethoxy)aniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$50.10

$50.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$50.10

About This Item

Linear Formula:
CF3OC6H4NH2
CAS Number:
Molecular Weight:
177.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

72-73 °C/8 mmHg (lit.)

density

1.325 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1cccc(OC(F)(F)F)c1

InChI

1S/C7H6F3NO/c8-7(9,10)12-6-3-1-2-5(11)4-6/h1-4H,11H2

InChI key

SADHVOSOZBAAGL-UHFFFAOYSA-N

General description

Synthesis of 3-(trifluoromethoxy)aniline from 2-chlorophenol has been reported.[1]

Application

3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A safe and economical synthesis of 3-(trifluoromethoxy) aniline from 2-chlorophenol.
Langlois B and Soula G.
Bulletin de la Societe Chimique De France, 6, 925-929 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service