Skip to Content
Merck
All Photos(1)

Key Documents

341266

Sigma-Aldrich

cyclic Isopropylidene 1,1-cyclopropanedicarboxylate

99%

Synonym(s):

1,1-Cyclopropanedicarboxylic acid cyclic isopropylidene ester, 6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$119.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$119.00

About This Item

Empirical Formula (Hill Notation):
C8H10O4
CAS Number:
Molecular Weight:
170.16
Beilstein:
1241835
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

99%

mp

63-65 °C (lit.)

solubility

DMF: soluble(lit.)
THF: soluble
diethyl ether: soluble(lit.)
water: insoluble(lit.)

functional group

ester
ketal

SMILES string

CC1(C)OC(=O)C2(CC2)C(=O)O1

InChI

1S/C8H10O4/c1-7(2)11-5(9)8(3-4-8)6(10)12-7/h3-4H2,1-2H3

InChI key

AXJVPXNVESYGDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service