Skip to Content
Merck
All Photos(3)

Key Documents

296988

Sigma-Aldrich

Octane

anhydrous, ≥99%

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696875
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
125-127 °C (lit.)
vapor pressure:
11 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

3.9 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

428 °F

expl. lim.

6.5 %

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.398 (lit.)

bp

125-127 °C (lit.)

mp

−57 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.703 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octane is linear alkane that can be synthesized from furfural, a renewable biomass resource. It can undergo catalytic cracking over alkali-treated MFI (Mordenite Framework Inverted) zeolites to mainly form primary cracking products.

Application


  • A photonic crystal material for the online detection of nonpolar hydrocarbon vapors.: Research on a novel photonic crystal material aimed at the efficient detection of nonpolar hydrocarbons such as octane, enhancing safety and monitoring in chemical processing environments (Bolshakov et al., 2022).


Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignition of non-premixed counterflow flames of octane and decane isomers.
Liu N, et al.
Proceedings of the Combustion Institute, 34(1), 903-910 (2013)
Nai-Tzu Chen et al.
Theranostics, 4(8), 798-807 (2014-06-24)
The unique optical properties of gold nanorods (GNRs) have recently drawn considerable interest from those working in in vivo biomolecular sensing and bioimaging. Especially appealing in these applications is the plasmon-enhanced photoluminescence of GNRs induced by two-photon excitation at infrared
Catalytic cracking of n-octane over alkali-treated MFI zeolites.
Jung JS, et al.
Applied Catalysis A: General, 288(1), 149-157 (2005)
Eagleson M.
Concise Encyclopedia Chemistry, 743-743 (1994)
Effective production of octane from biomass derivatives under mild conditions.
Xu W, et al.
ChemSusChem, 4(12), 1758-1761 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service