Skip to Content
Merck
All Photos(1)

Key Documents

338672

Sigma-Aldrich

4′-(Octyloxy)-4-biphenylcarbonitrile

liquid crystal (nematic)

Synonym(s):

4′-(n -Octyloxy)-4-cyanobiphenyl, 4′-(Octyloxy)-4-biphenylcarbonitrile, 4′-(Octyloxy)-4-cyanobiphenyl, 4′-(Octyloxy)[1,1′-biphenyl]-4-carbonitrile, 4′-Octyloxy-1,1′-biphenyl-4-carbonitrile, 4-(4-Octyloxyphenyl)benzonitrile, 4-(Octyloxy)-4′-cyanobiphenyl, 4-Cyano-4′-(n -octyloxy)biphenyl, 4-Cyano-4′-(octyloxy)-1,1′-biphenyl, 4-Cyano-4′-(octyloxy)biphenyl, 4-Cyano-4′-n -octyloxy-1,1′-biphenyl, p -(Octyloxy)-p ′-cyanobiphenyl, p -Cyano-p ′-(octyloxy)biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OC6H4C6H4CN
CAS Number:
Molecular Weight:
307.43
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid crystal (nematic)

mp

51-77 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C21H25NO/c1-2-3-4-5-6-7-16-23-21-14-12-20(13-15-21)19-10-8-18(17-22)9-11-19/h8-15H,2-7,16H2,1H3

InChI key

GPGGNNIMKOVSAG-UHFFFAOYSA-N

General description

4′-octyl-4- biphenylcarbonitrile (8CB) is a low molar mass liquid crystal (LCs), which shows different liquid crystalline phases depending on temperature conditions. Under normal temperature conditions 8CB exists only as bulk smectic-A phase.[1] LCs are responsive to minor changes in temperature, shear, electric (or magnetic) fields, and the structure of solid surfaces with which they are in contact.[2]

Application

8CB may be used in the preparation of asymmetrical zinc phthalocyanine (aZnPc)-functional photocurable copolymer.[3]
Monolayer films of the LCs deposited onto highly ordered pyrolytic graphite, these self-assembled films display a wide variety of structural forms.[4]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of molecular ordering in monolayer liquid crystal films.
Patrick DL, et al.
The Journal of Physical Chemistry, 100(20), 8478-8481 (1996)
Preparation of a new polymer-dispersed liquid crystal film by using phthalocyanine-functional photocurable copolymer.
Mert H, et al.
Journal of Applied Polymer Science, 132(10) (2015)
Substrate defects and variations in interfacial ordering of monolayer molecular films on graphite.
Patrick DL, et al.
Langmuir, 10(1), 298-302 (1994)
Separation of nematic and smectic-A potential effects on solute ordering in a series of binary 6OCB-8OCB mixtures
Burnell EE, et al.
Canadian Journal of Chemistry, 89(7), 900-908 (2011)

Questions

  1. Give the phase transition temperatures and mention the different phases of liquid crystalline compound 338672 4'-(octyloxy)-4-biphenyl carbonitrile

    1 answer
    1. The phase transition temperatures for this compound have not been determined. See the link below to the PubChem file for this material. There are a number of references and physical properties listed that may be of interest:
      https://pubchem.ncbi.nlm.nih.gov/compound/104173

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service