Skip to Content
Merck
All Photos(1)

Key Documents

588474

Sigma-Aldrich

6-[4-(4-Cyanophenyl)phenoxy]hexyl methacrylate

96%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$423.00

$423.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$423.00

About This Item

Empirical Formula (Hill Notation):
C23H25NO3
CAS Number:
Molecular Weight:
363.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$423.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

96%

form

liquid crystal

mp

73-76 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCCCCCOc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C23H25NO3/c1-18(2)23(25)27-16-6-4-3-5-15-26-22-13-11-21(12-14-22)20-9-7-19(17-24)8-10-20/h7-14H,1,3-6,15-16H2,2H3

InChI key

JJJGWRPDCQICFP-UHFFFAOYSA-N

Application

Used in the synthesis of side chain liquid crystalline homopolymers and block copolymers with the cyanobiphenyl group as the mesogen.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service