Skip to Content
Merck
All Photos(1)

Key Documents

338389

Sigma-Aldrich

Triphenylphosphine hydrobromide

97%

Synonym(s):

Ph3P · HBr, Ph3P · HBr, Triphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P · HBr
CAS Number:
Molecular Weight:
343.20
Beilstein:
3633387
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

196 °C (dec.) (lit.)

functional group

phosphine

SMILES string

Br.c1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C18H15P.BrH/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h1-15H;1H

InChI key

CMSYDJVRTHCWFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as mild source of anhydrous HBr; catalyst for formation of THP ethers from tertiary alcohols; preparation of phosphonium salts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hamanaka, N.; Kosuge, S.; Iguchi, S.
Synlett, 139-139 (1990)
Kaila, N.; Blumenstein, M. et al.
The Journal of Organic Chemistry, 57, 4576-4576 (1992)
Tong Wang et al.
Organic & biomolecular chemistry, 9(14), 5260-5265 (2011-06-02)
The triphenylphosphine-catalyzed formal [3 + 2] cycloaddition of allenoates and trifluoromethylketones was realized to give the corresponding dihydrofurans in good yields with excellent γ-regioselectivities. Hydrogenation of the dihydrofurans gave 2,4,4-trisubstituted tetrahydrofurans in good yields with exclusive cis-selectivities.
Yong Ma et al.
Langmuir : the ACS journal of surfaces and colloids, 27(21), 13097-13103 (2011-09-21)
Chemically selective liposomal surface functionalization and liposomal microarray fabrication using azide-reactive liposomes are described. First, liposome carrying PEG-triphenylphosphine was prepared for Staudinger ligation with azide-containing biotin, which was conducted in PBS buffer (pH 7.4) at room temperature without a catalyst.
Kirill Nikitin et al.
Chemical communications (Cambridge, England), 49(14), 1434-1436 (2013-01-17)
Triphenylhalophosphonium halides, Ph(3)PX(2), form crystals comprising bridged linear cations [Ph(3)P-X-X-X-PPh(3)](+) where the X(3) bridge is shortened from 6.56 Å in Cl-Cl-Cl to 6.37 Å in the Br-Br-Br system. It is proposed that this structure is stabilised by five-centre/six-electron (5c-6e) hypervalent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service