Skip to Content
Merck
All Photos(1)

Documents

338303

Sigma-Aldrich

Chloroethane solution

2.0 M in tert-butyl methyl ether, anhydrous

Synonym(s):

Ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050105
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

vapor density

2.22 (vs air)

vapor pressure

20.03 psi ( 55 °C)
5.98 psi ( 20 °C)

form

liquid

autoignition temp.

966 °F

concentration

2.0 M in tert-butyl methyl ether

impurities

<0.005% water

density

0.775 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroethane is an alkyl halide. Chloroethane (Ethyl chloride) adsorbed on Ag(111) at 100K has been studied using temperature programmed desorption, work function change, ultraviolet and X-ray photoelectron spectroscopy.

Application

  • Green solvent in biomaterial extraction: Research highlights the use of Chloroethane as an effective solvent for extracting lignin-containing nanocellulose fibrils from date palm waste, emphasizing its potential in enhancing the sustainability of biomaterial processing (Raza et al., 2024).

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:
For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkyl halide photochemistry on Ag (111): I. Ethyl chloride.
Zhou X-L and White JM.
Surface Science, 241(3), 244-258 (1991)
Suzuko Yamazaki et al.
The journal of physical chemistry. A, 114(15), 5092-5098 (2010-03-27)
The photocatalytic degradation of chlorinated ethanes was studied in a tubular photoreactor packed with TiO(2) pellets prepared by sol-gel method. The steady-state condition was not obtained, but the deterioration in the photocatalytic activity was observed during the irradiation. Effects of
Peter Barnett
Pediatric emergency care, 25(6), 415-419 (2009-06-17)
Conscious sedation is used frequently to perform procedures that may be undertaken with or without minimal sedation. Fracture manipulation can be performed with minimal discomfort in the awake patient using various techniques-for example, intravenous regional anesthesia, nerve blocks, and hematoma
Xi-Guang Wei et al.
The Journal of organic chemistry, 75(12), 4212-4217 (2010-05-21)
The enhanced reactivity exhibited by six pseudo-alpha-bases, RC[triple bond]CZ(-) (R = H and Cl; Z = O, S, and Se) in gas-phase E2 reactions with ethyl chloride was examined at the G2(+) level. It is found that anomalous reactivity is
Lindsey L Cohen et al.
The Clinical journal of pain, 25(6), 490-494 (2009-06-23)
The purpose of this study was to evaluate the effectiveness of vapocoolant for preschoolers' immunization injection pain relief. Fifty-seven 4 to 6-year-old children were randomized into vapocoolant alone or typical care conditions. Pain was measured at the baseline and at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service