Skip to Content
Merck
All Photos(1)

Key Documents

338273

Sigma-Aldrich

2-Methylhexanoic acid

99%

Synonym(s):

2-Methylcaproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$47.10

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$47.10

About This Item

Linear Formula:
CH3(CH2)3CH(CH3)COOH
CAS Number:
Molecular Weight:
130.18
Beilstein:
1721227
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

0.0576 mmHg ( 25 °C)(lit.)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.422 (lit.)

bp

209-210 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCC(C)C(O)=O

InChI

1S/C7H14O2/c1-3-4-5-6(2)7(8)9/h6H,3-5H2,1-2H3,(H,8,9)

InChI key

CVKMFSAVYPAZTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fawzia H Abdel-Rahman et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 43(8), 680-685 (2008-10-23)
Thirty-three organic acids and furfural metabolites were examined for their nematicidal activity against plant-parasitic, free-living and predacious nematodes. Propionic acid, 2-methylhexanoic acid, lactic acid, maleic acid, and furic acid were the most effective nematicides among normal chain organic acids, branched
J Julák et al.
Medical science monitor : international medical journal of experimental and clinical research, 6(3), 605-610 (2001-02-24)
The volatile fatty acid profiles were determined by a simple gas chromatographic method in 375 microbiologically positive and negative blood cultures. Aerobic bacteria yielded profiles with low content of acids, some of which were chromatographically negative. Anaerobic bacteria produced more
How is chemical interesterification initiated: Nucleophilic substitution or a-proton abstraction?.
Liu L.
Journal of the American Oil Chemists' Society, 81(4), 331-337 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service