Skip to Content
Merck
All Photos(2)

Documents

335428

Sigma-Aldrich

4,4′-Dinitro-2-biphenylamine

98%

Synonym(s):

2-(4-Nitrophenyl)-5-nitroaniline, 4,4′-Dinitro-[1,1′-biphenyl]-2-amine, 5-nitro-2-(4-nitrophenyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4C6H3(NO2)NH2
CAS Number:
Molecular Weight:
259.22
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

98%

form

powder, crystals or chunks

mp

208-210 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1cc(ccc1-c2ccc(cc2)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C12H9N3O4/c13-12-7-10(15(18)19)5-6-11(12)8-1-3-9(4-2-8)14(16)17/h1-7H,13H2

InChI key

FRZPYFUNNLIMEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Dinitro-2-biphenylamine is a nitrated aromatic amine that is a direct mutagen. In Salmonella typhimurium TA100, 4,4′-dinitro-2-biphenylamine causes base-pair reversion in the histidine locus.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutagenicity and antimutagenicity studies of tannic acid and its related compounds.
Chen SC and Chung KT.
Food And Chemical Toxicology, 38(1), 1-5 (2000)
S C Chen et al.
Mutation research, 395(2-3), 223-227 (1998-02-18)
Among p-phenylenediamine, benzidine and the analogues we previously tested, only the nitro-group containing 2-nitro-p-phenylenediamine, 3-nitro-o-phenylenediamine, 4-nitro-o-phenylenediamine and 4,4'-dinitro-2-biphenylamine caused base-pair reversion in the histidine locus of Salmonella typhimurium TA100. In order to determine the types of mutations involved, such as
K Chung et al.
Mutation research, 465(1-2), 165-171 (2000-03-10)
Four nitrated aromatic amines (2-nitro-p-phenylenediamine [2NPD], 3-nitro-o-phenylenediamine [3NPD], 4-nitro-o-phenylenediamine [4NPD] and 4,4'-dinitro-2-biphenylamine [DNBA]) are direct-acting mutagens in Salmonella typhimurium strain TA100. These compounds were tested further using the Xenometrix strains of S. typhimurium: TA7001, TA7002, TA7003, TA7004, TA7005, and TA7006

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service