Skip to Content
Merck
All Photos(2)

Key Documents

333360

Sigma-Aldrich

Boron

crystalline, −60 mesh, 99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
EC Number:
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99% trace metals basis

form

crystalline

resistivity

1.5E12 μΩ-cm, 20 °C

impurities

<0.5% carbon

particle size

−60 mesh

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel L Silverio et al.
Nature, 494(7436), 216-221 (2013-02-15)
The discovery of catalysts that can be used to synthesize complex organic compounds by enantioselective transformations is central to advances in the life sciences; for this reason, many chemists aim to discover catalysts that allow for preparation of chiral molecules
Tara A Devirian et al.
Critical reviews in food science and nutrition, 43(2), 219-231 (2003-04-23)
Boron may be an essential nutrient for animals and humans. Dietary boron influences the activity of many metabolic enzymes, as well as the metabolism of steroid hormones and several micronutrients, including calcium, magnesium, and vitamin D. Boron supplementation in rats
Yu Zhao et al.
Journal of the American Chemical Society, 135(4), 1201-1204 (2013-01-16)
Two kinds of boron and nitrogen co-doped carbon nanotubes (CNTs) dominated by bonded or separated B and N are intentionally prepared, which present distinct oxygen reduction reaction (ORR) performances. The experimental and theoretical results indicate that the bonded case cannot
Boron containing compounds as protease inhibitors.
Reem Smoum et al.
Chemical reviews, 112(7), 4156-4220 (2012-04-24)
Anne-Marie Alexander et al.
Chemical Society reviews, 39(11), 4388-4401 (2010-06-08)
Catalysts generated by the addition of carbon, nitrogen or phosphorus to transition metals have interesting properties and potential applications. The addition of carbon, nitrogen or phosphorus can lead to substantial modification of the catalytic efficacy of the parent metal and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service