Skip to Content
Merck
All Photos(1)

Documents

33240

Sigma-Aldrich

2,6-Diaminopimelic acid

≥95.0% (NT), for peptide synthesis

Synonym(s):

2,6-Diaminoheptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(NH2)(CH2)3CH(NH2)COOH
CAS Number:
Molecular Weight:
190.20
Beilstein:
1787719
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

product name

2,6-Diaminopimelic acid, ≥95.0% (NT)

Assay

≥95.0% (NT)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

~295 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(CCCC(N)C(O)=O)C(O)=O

InChI

1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)

InChI key

GMKMEZVLHJARHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Diaminopimelic acid (2,6-DAP) can be used as a building block in the synthesis of tripeptides , and γ-glutamyldiaminopimelic acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new stereocontrolled synthesis of uncommon tripeptides derived from 2, 6-diaminopimelic acid (2, 6-DAP)
Paradisi F, et al.
Tetrahedron Asymmetry, 12(23), 3319-3324 (2001)
Peter Kämpfer et al.
International journal of systematic and evolutionary microbiology, 64(Pt 5), 1811-1816 (2014-02-22)
A Gram-staining-positive, aerobic, non-endospore forming organism, isolated as a seed endophyte (colonizing the internal healthy tissue of plant seed) of sweet corn (Zea mays), strain CSE-5610T, was studied for its taxonomic allocation. On the basis of 16S rRNA gene sequence
Sarah N Buss et al.
International journal of systematic and evolutionary microbiology, 63(Pt 11), 4087-4093 (2013-06-04)
A Gram-staining-positive, endospore-forming rod was isolated independently from clinical specimens in New York State, USA, once in 2009 and twice in 2011. The three isolates had identical 16S rRNA gene sequences and, based on their 16S rRNA gene sequence, are
Lilian Hor et al.
Biochimie, 95(10), 1949-1953 (2013-07-11)
DAP epimerase is the penultimate enzyme in the lysine biosynthesis pathway. The most versatile assay for DAP epimerase catalytic activity employs a coupled DAP epimerase-DAP dehydrogenase enzyme system with a commercial mixture of DAP isomers as substrate. DAP dehydrogenase converts
Structure- activity relationships in nucleotide oligomerization domain 1 (Nod1) agonistic γ-glutamyldiaminopimelic acid derivatives
Agnihotri G, et al.
Journal of Medicinal Chemistry, 54(5), 1490-1510 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service