Skip to Content
Merck
All Photos(1)

Documents

329010

Sigma-Aldrich

Gallium arsenide

pieces, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaAs
CAS Number:
Molecular Weight:
144.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999% trace metals basis

form

pieces

density

5.31 g/mL at 25 °C (lit.)

SMILES string

[Ga]#[As]

InChI

1S/As.Ga

InChI key

JBRZTFJDHDCESZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 1B - STOT RE 1

Target Organs

Respiratory system,hematopoietic system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Wei Tan et al.
PloS one, 7(11), e50681-e50681 (2012-12-12)
Characterization and modeling of metal-semiconductor-metal (MSM) GaAs diodes using to evaporate SiO₂ and Pd simultaneously as a mixture electrode (called M-MSM diodes) compared with similar to evaporate Pd as the electrode (called Pd-MSM diodes) were reported. The barrier height (φ(b))
Yu Bomze et al.
Physical review letters, 109(2), 026801-026801 (2012-10-04)
We report on measurements of first-passage-time distributions associated with current switching in weakly coupled GaAs/AlAs superlattices driven by shot noise, a system that is both far from equilibrium and high dimensional. Static current-voltage (I-V) characteristics exhibit multiple current branches and
Chao-Wei Hsu et al.
Nanotechnology, 23(49), 495306-495306 (2012-11-17)
GaAs is grown by metal-organic vapor-phase epitaxy on a 55 nm round-hole patterned Si substrate with SiO(2) as a mask. The threading dislocations, which are stacked on the lowest energy facet plane, move along the SiO(2) walls, reducing the number
I I Yakimenko et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 072201-072201 (2013-01-19)
We analyze the occurrence of local magnetization and the effects of electron localization in different models of quantum point contacts (QPCs) using spin-relaxed density functional theory (DFT/LSDA) by means of numerical simulations. In the case of soft confinement potentials the
M Baranowski et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(6), 065801-065801 (2013-01-12)
In this study we apply time resolved photoluminescence and contactless electroreflectance to study the carrier collection efficiency of a GaInNAsSb/GaAs quantum well (QW). We show that the enhancement of photoluminescence from GaInNAsSb quantum wells annealed at different temperatures originates not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service