Skip to Content
Merck
All Photos(1)

Key Documents

328529

Sigma-Aldrich

4′-(Pentyloxy)-4-biphenylcarbonitrile

liquid crystal (nematic), 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OC6H4C6H4CN
CAS Number:
Molecular Weight:
265.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid crystal (nematic)

SMILES string

CCCCCOc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C18H19NO/c1-2-3-4-13-20-18-11-9-17(10-12-18)16-7-5-15(14-19)6-8-16/h5-12H,2-4,13H2,1H3

InChI key

RDISTOCQRJJICR-UHFFFAOYSA-N

General description

4′-(Pentyloxy)-4-biphenylcarbonitrile (5-OCB) liquid crystals are the member of the oxycyanobiphenyl family.[1] It exhibits liquid crystalline phase in the temperature range of 47-67.5oC.[1] Crystal and molecular structure of 5-OCB was reported. The crystals belong to the monoclinic system (a=21.378, b=5.695, c=12.789Å).[2] The 5-OCB molecules are antiparallel in layers. H NMR of the phase transitions has been studied.[1]

Application

5-OCB may be used in indicators and storage devices.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-Ray Studies on the Mesogen 4?-n-Pentyloxy-4-Biphenylcarbonitrile(5OCB) in the Solid Crystalline State
Mandal P and Paul S
Mol. Cryst. Liq. Cryst., 131(3-4) (1985)
Wide-line 1H NMR study of 4?-(pentyloxy)-4-biphenylcarbonitrile in different phase states
Chuvaev VF, et al.
Russian Journal of Inorganic Chemistry, 53(12), 1925-1928 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service