Skip to Content
Merck
All Photos(1)

Documents

326216

Sigma-Aldrich

Silver pentafluoropropionate

98%

Synonym(s):

Pentafluoropropionic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2F5CO2Ag
CAS Number:
Molecular Weight:
271.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

242-244 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(=O)O[Ag]

InChI

1S/C3HF5O2.Ag/c4-2(5,1(9)10)3(6,7)8;/h(H,9,10);/q;+1/p-1

InChI key

XAYJXAUUXJTOSI-UHFFFAOYSA-M

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H B Lee et al.
Journal of AOAC International, 81(6), 1209-1216 (1998-12-16)
The paper describes a simple and quantitative method for monitoring non-conjugated 17 beta-estradiol (E2) and its metabolites estrone (E1) and estriol (E3) as environmental contaminants in municipal sewage effluents. Estrogens were preconcentrated and cleaned up by solid-phase extraction using a
B Li et al.
Journal of pharmaceutical and biomedical analysis, 55(1), 78-84 (2011-02-15)
Liquid chromatography combined with pulsed electrochemical detection (LC-PED) is the method of choice in the European Pharmacopoeia for the determination of gentamicin and its related substances. A recently approved improved LC-PED method, with a reversed-phase C(18) column and a mobile
Sandra Rinne et al.
Journal of chromatography. A, 1119(1-2), 285-293 (2006-03-21)
A fast and sensitive capillary liquid chromatography (cLC) column-switching method with electrospray ionization time-of-flight mass spectrometry (ESI-TOF-MS) detection for the simultaneous determination of dopamine (D), epinephrine (E), norepinephrine (NE) and serotonin (SE) was pursued. A sample volume of 100 microl
[Recent development of C-terminal sequence analysis of proteins].
A Tsugita
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 43(7), 857-865 (1998-06-23)
Sabina Strano-Rossi et al.
Journal of applied toxicology : JAT, 31(7), 649-654 (2010-12-07)
A rapid and sensitive method for the simultaneous determination of alfentanyl, sufentanyl and fentanyl (and its major metabolite norfentanyl) in urine was developed and validated. The method involved a liquid-liquid extraction in alkaline conditions, derivatization with pentafluoropropionic anhydride to improve

Articles

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service