Skip to Content
Merck
All Photos(1)

Documents

325228

Sigma-Aldrich

2-Methylpyridine-3-carboxylic acid

98%

Synonym(s):

2-Methylnicotinic acid, 2-Methylpyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
114333
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

228-230 °C (dec.) (lit.)

SMILES string

Cc1ncccc1C(O)=O

InChI

1S/C7H7NO2/c1-5-6(7(9)10)3-2-4-8-5/h2-4H,1H3,(H,9,10)

InChI key

HNTZKNJGAFJMHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylpyridine-3-carboxylic acid was used in synthesis of 1,8-dioxo-1,2,7,8-tetrahydro-2,7,10-triaza-anthracene-4,5-dicarbaldehydes (DOTTADs) and their imines. It was also used in synthesis of 7,7-dichloro-5,7-dihydro-thieno[3,4-b]pyridin-5-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwini Kumar Ray et al.
Molecular and biochemical parasitology, 219, 42-51 (2017-11-28)
Selective 2'-hydroxyl acylation analyzed by primer extension (SHAPE) is a versatile sequence independent method to probe RNA structure in vivo and in vitro. It has so far been tried mainly with model organisms. We show that cells of Entamoeba histolytica
Andrea Arany et al.
Organic & biomolecular chemistry, 1(9), 1545-1551 (2003-08-21)
The interaction of Hantzsch pyridinecarboxylic acids with dialkylformamides and POCl3, followed by treatment with NH4OH yields 1,8-dioxo-1,2,7,8-tetrahydro-2,7,10-triazaanthracenes (DOTTADs), which have great potential as useful ligands for Group I and II metals and some transition metals. The corresponding Hantszch esters similarly
Preparation, X-ray structure and propylaminolysis of 7, 7-dichloro-5, 7-dihydro-thieno [3, 4-b] pyridin-5-one.
van Es T, et al.
J. Chem. Res. (M), 2007(6), 373-376 (2007)
Ignaz J Buerge et al.
Environmental science & technology, 53(10), 5725-5732 (2019-04-25)
Many pesticides show a pronounced biphasic degradation in soil, typically with a faster initial phase, followed by a slower decline. For chiral compounds, a biphasic decline of the total concentration may result from enantioselective degradation. In this study with the
Naoki Takizawa et al.
Virology, 531, 126-140 (2019-03-16)
The influenza A virus genome consists of eight single-stranded negative-sense RNA segments. The noncoding regions located at the 3'- and 5'- ends of each segment are necessary for genome packaging, and the terminal coding regions are required to precisely bundle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service