![2-Methylpyridine-3-carboxylic acid 98%](/deepweb/assets/sigmaaldrich/product/structures/853/063/1fda3859-cdf7-4922-a297-5a645b803188/640/1fda3859-cdf7-4922-a297-5a645b803188.png)
2-Methylpyridine-3-carboxylic acid
![2-Methylpyridine-3-carboxylic acid 98%](/deepweb/assets/sigmaaldrich/product/structures/853/063/1fda3859-cdf7-4922-a297-5a645b803188/640/1fda3859-cdf7-4922-a297-5a645b803188.png)
Synonym(s):
2-Methylnicotinic acid, 2-Methylpyridine-3-carboxylic acid
Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing