Skip to Content
Merck
All Photos(1)

Key Documents

324752

Sigma-Aldrich

1,2,3-Trifluorobenzene

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$40.40

$40.40


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$40.40

About This Item

Empirical Formula (Hill Notation):
C6H3F3
CAS Number:
Molecular Weight:
132.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.40


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

≥98%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

94-95 °C (lit.)

density

1.28 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1cccc(F)c1F

InChI

1S/C6H3F3/c7-4-2-1-3-5(8)6(4)9/h1-3H

InChI key

AJKNNUJQFALRIK-UHFFFAOYSA-N

General description

Crystal structure 1,2,3-trifluorobenzene was reported.[1] The microwave spectrum of 1,2,3-trifluorobenzene was studied using a molecular beam Fourier transform microwave spectrometer.[2] Laser-induced fluorescence spectra of the radical cation of 1,2,3-trifluorobenzene was determined in both the gas phase and solid Ne matrices.[3]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael T Kirchner et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 11), o2670-o2670 (2009-01-01)
In the title compound, C(6)H(3)F(3), weak electrostatic and dispersive forces between C(δ+)-F(δ-) and H(δ+)-C(δ-) groups are at the borderline of the hydrogen-bond phenomenon and are poorly directional and further deformed in the presence of π-π stacking inter-actions. The mol-ecule lies
Marissa A Dobulis et al.
The Journal of chemical physics, 152(20), 204309-204309 (2020-06-04)
The broadband photoelectron source realized by detaching O2-·X (X = neutral unsaturated molecule) complexes offers a unique opportunity to probe temporary anion states of the unsaturated species. Detachment of the ion molecule complex typically accesses a dissociative portion of the
Microwave Spectrum of 1, 2, 3-Trifluorobenzene.
Onda M, et al.
Journal of Molecular Spectroscopy, 169(2), 480-483 (1995)
The laser-induced fluorescence spectrum of the 1, 2, 3-trifluorobenzene radical cation.
Bondybey VE, et al.
Journal of Molecular Spectroscopy, 84(1), 124-131 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service