Skip to Content
Merck
All Photos(1)

Key Documents

32008

Sigma-Aldrich

1,2-Diethylbenzene

≥99.0% (GC)

Synonym(s):

o-Diethylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$160.00
5 ML
$583.00

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$160.00
5 ML
$583.00

About This Item

Linear Formula:
C6H4(C2H5)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1904392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (GC)

autoignition temp.

743 °F

refractive index

n20/D 1.502 (lit.)
n20/D 1.503

bp

183 °C (lit.)

mp

−31 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1CC

InChI

1S/C10H14/c1-3-9-7-5-6-8-10(9)4-2/h5-8H,3-4H2,1-2H3

InChI key

KVNYFPKFSJIPBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Diethylbenzene can be used as a reactant in the preparation of pseudocyclic diaryliodonium triflates, 5,6-disubstituted indanone intermediates via intramolecular Friedel-Crafts alkylation, dibromide intermediates via radical bromination, and in the dehydrogenative C-H/C-H arylation of indolines[1][2][3][4].

Application

  • Attraction of adult Harmonia axyridis to volatiles of the insectary plant Cnidium monnieri: This study investigates the attraction of the beetle Harmonia axyridis to 1,2-diethylbenzene, indicating potential uses in biological control strategies (Zhiping et al., 2020).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of 4, 5, 6, 7-tetrahydroisoindoles by palladium-catalyzed hydride reduction
Hou D, et al.
The Journal of Organic Chemistry, 72 (2007)
Synthesis of arylbenziodoxoles using pseudocyclic benziodoxole triflate and arenes
Yoshimura A, et al.
ARKIVOC (Gainesville, FL, United States), 2020 (2021)
Exceptionally Mild Palladium (II)-Catalyzed Dehydrogenative C--H/C--H Arylation of Indolines at the C-7 Position under Air
Jiao L, et al.
Organic Letters, 16 (2014)
Jean-Paul Payan et al.
Archives of toxicology, 82(9), 591-600 (2008-02-07)
The bio-distribution of the neurotoxic 1,2-diethylbenzene (1,2-DEB) was studied in male Sprague-Dawley rats after intravenous administration of [(14)C] 1,2-DEB (1 mg kg(-1)). The highest concentrations of [(14)C] non-volatile metabolites, determined by whole-body auto-radiography, were in the nasal cavity, ethmoid turbinates
Desire Tshala-Katumbay et al.
Toxicological sciences : an official journal of the Society of Toxicology, 105(1), 134-141 (2008-05-27)
Motor neuron axonopathy in diseases such as amyotrophic lateral sclerosis can be modeled and probed with neurotoxic chemicals that induce similar patterns of pathology, such as axonal spheroids that represent focal accumulation of anterogradely transported neurofilaments (NFs). The aromatic gamma-diketone-like

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service