Skip to Content
Merck
All Photos(1)

Key Documents

319104

Sigma-Aldrich

Methyl Red sodium salt solution

0.1 wt. % in H2O

Synonym(s):

2-[4-(Dimethylamino)phenylazo]benzoic acid sodium salt, 4-Dimethylaminoazobenzene-2′-carboxylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4CO2Na
CAS Number:
Molecular Weight:
291.28
Beilstein:
4065488
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

concentration

0.1 wt. % in H2O

pH

4.2-6.2, pink to yellow

density

0.989 g/mL at 25 °C

λmax

437 nm

SMILES string

[Na+].CN(C)c1ccc(cc1)\N=N\c2ccccc2C([O-])=O

InChI

1S/C15H15N3O2.Na/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);/q;+1/p-1/b17-16+;

InChI key

GNTPCYMJCJNRQB-CMBBICFISA-M

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Danielle Kelley et al.
Biochemistry, 49(26), 5405-5407 (2010-06-15)
It is shown that Methyl Red can be used as an indicator dye that changes color in Escherichia coli culture as a result of time- and cell density-dependent bleaching by azoreductase produced by the bacteria. For cell cultures that are
Oana Alexandru et al.
Journal of neuro-oncology, 102(1), 9-18 (2010-07-17)
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of
Jinhui Feng et al.
Archives of biochemistry and biophysics, 520(2), 99-107 (2012-03-06)
AzoA from Enterococcus faecalis is a member of the polymeric flavin-dependent NADH-preferred azoreductase group. Little is known about the binding and interaction of NADH and azo dye in the azoreductase group. A synergetic strategy based on computational prediction, reverse genetics
Lamia Ayed et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 62(12), 2837-2845 (2010-12-03)
With the aid of analysis software (Minitab 14.0), the formulation of pure culture in Mineral Salts Medium (MSM) can be optimized for several responses and the best formulation can be obtained. The influence of the different mixtures of three strains
X D Zhang et al.
Journal of hazardous materials, 170(2-3), 883-887 (2009-06-09)
An aqueous C.I. Acid Red 2 solution was decolorized by electrolysis using iron as anode. The decolorization mechanism was investigated through experimental observations on the electrochemical behavior of C.I. Acid Red 2 on Pt rotating disk electrode, UV-visible spectra of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service