Skip to Content
Merck
All Photos(3)

Key Documents

114502

Sigma-Aldrich

Methyl Red sodium salt

ACS reagent, Dye content 95 %

Synonym(s):

4-Dimethylaminoazobenzene-2′-carboxylic acid sodium salt, Acid Red 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$88.10
100 G
$267.00

$88.10


Estimated to ship onApril 07, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$88.10
100 G
$267.00

About This Item

Empirical Formula (Hill Notation):
C15H14N3NaO2
CAS Number:
Molecular Weight:
291.28
Colour Index Number:
13020
Beilstein:
4065488
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$88.10


Estimated to ship onApril 07, 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

form

powder

composition

Dye content, 95%

technique(s)

titration: suitable

visual transition interval

4.2-6.2, pink to yellow

solubility

water: 0.1%

λmax

437 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CN(C)c1ccc(cc1)\N=N\c2ccccc2C([O-])=O

InChI

1S/C15H15N3O2.Na/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);/q;+1/p-1/b17-16+;

InChI key

GNTPCYMJCJNRQB-CMBBICFISA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Methyl Red is a pH indicator and it changes color at a pH of 5.5.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved dithizone method for the determination of small quantities of zinc in blood and tissue samples.
Vallee BL and Gibson JG 2nd
The Journal of Biological Chemistry, 176, 435-443 (1948)
Reza Amin et al.
Micromachines, 11(6) (2020-07-01)
To transform from reactive to proactive healthcare, there is an increasing need for low-cost and portable assays to continuously perform health measurements. The paper-based analytical devices could be a potential fit for this need. To miniaturize the multiplex paper-based microfluidic
Jinhui Feng et al.
Archives of biochemistry and biophysics, 520(2), 99-107 (2012-03-06)
AzoA from Enterococcus faecalis is a member of the polymeric flavin-dependent NADH-preferred azoreductase group. Little is known about the binding and interaction of NADH and azo dye in the azoreductase group. A synergetic strategy based on computational prediction, reverse genetics
X D Zhang et al.
Journal of hazardous materials, 170(2-3), 883-887 (2009-06-09)
An aqueous C.I. Acid Red 2 solution was decolorized by electrolysis using iron as anode. The decolorization mechanism was investigated through experimental observations on the electrochemical behavior of C.I. Acid Red 2 on Pt rotating disk electrode, UV-visible spectra of
Robin L Stingley et al.
Journal of medical microbiology, 59(Pt 1), 108-114 (2009-09-05)
Reduction of Methyl Red (MR) and Orange II (Or II) by 26 human skin bacterial species was monitored by a rapid spectrophotometric assay. The analysis indicated that skin bacteria, representing the genera Staphylococcus, Corynebacterium, Micrococcus, Dermacoccus and Kocuria, were able

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service