Skip to Content
Merck
All Photos(1)

Key Documents

318043

Sigma-Aldrich

Difluoromethane

99.7%

Synonym(s):

HFC-32

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2F2
CAS Number:
Molecular Weight:
52.02
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99.7%

bp

−51.6 °C (lit.)

mp

−136 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FCF

InChI

1S/CH2F2/c2-1-3/h1H2

InChI key

RWRIWBAIICGTTQ-UHFFFAOYSA-N

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bifurcated CH2 ... O and (C-H)2 ... F-C weak hydrogen bonds: the oxirane-difluoromethane complex.
Susana Blanco et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 5(11), 1779-1782 (2004-12-08)
Jason M Keller et al.
Environmental science & technology, 37(14), 3141-3144 (2003-08-07)
Field-scale tests were performed to evaluate the effectiveness of the gas-phase partitioning tracer method for in-situ measurement of soil-water content. The tracer tests were conducted before and after a controlled infiltration event to evaluate performance at two water contents. Nonpartitioning
Titus V Albu et al.
Journal of molecular modeling, 13(11), 1109-1121 (2007-08-21)
Accurate potential energy surfaces for the OH + CH2F2 --> H2O + CHF2 reaction are constructed using hybrid and hybrid meta density functional theory methods (mPW1PW91, B1B95, and mPW1B95) with specific reaction parameters in conjunction with the 6-31 + G(d,p)
Heonki Kim et al.
Environmental science & technology, 41(1), 235-241 (2007-02-03)
In this laboratory study, a new experimental method involving the use of a set of four gaseous tracers, was developed for measuring the NAPL saturation directly accessible to the mobile gas as well as the total NAPL saturation in unsaturated
M K Ellis et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 31(2), 243-251 (1996-06-01)
Difluoromethane (HFC32) is under development as a replacement for chlorofluorocarbons (CFCs) in some refrigeration applications. It has been evaluated by standard studies of toxicity, developmental toxicity, and genotoxicity. In addition, the metabolism and disposition of HFC32 was investigated and a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service