Skip to Content
Merck
All Photos(1)

Key Documents

310891

Sigma-Aldrich

10-Bromo-1-decanol

technical grade, 90%

Synonym(s):

Decamethylene bromohydrin

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$140.00

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$140.00

About This Item

Linear Formula:
Br(CH2)10OH
CAS Number:
Molecular Weight:
237.18
Beilstein:
1738593
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.476 (lit.)

bp

165-169 °C/10 mmHg (lit.)

density

1.19 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

OCCCCCCCCCCBr

InChI

1S/C10H21BrO/c11-9-7-5-3-1-2-4-6-8-10-12/h12H,1-10H2

InChI key

LGZMUUBPTDRQQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

10-Bromo-1-decanol was used in the synthesis of (11Z,13Z)-11,13-hexadecadienal, the female sex pheromone of the navel orangeworm (Amyelois transitella).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Mori
Bioscience, biotechnology, and biochemistry, 73(12), 2727-2730 (2009-12-08)
(11Z,13Z)-11,13-hexadecadienal, the female sex pheromone of the navel orangeworm (Amyelois transitella), was synthesized from commercially available 10-bromo-1-decanol in a 27% overall yield (8 steps). The synthesis was achieved by using 10-iododecanal, trimethylsilylacetylene and (Z)-1-bromo-1-butene as the key building blocks, employing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service