Skip to Content
Merck
All Photos(1)

Key Documents

305782

Sigma-Aldrich

4-(3-Oxobutyl)phenyl acetate

≥96%

Synonym(s):

4-(4-Acetoxyphenyl)-2-butanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$49.70

$49.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$49.70

About This Item

Linear Formula:
CH3CO2C6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
206.24
Beilstein:
1961620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$49.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥96%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

123-124 °C/0.2 mmHg (lit.)

density

1.099 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CC(=O)CCc1ccc(OC(C)=O)cc1

InChI

1S/C12H14O3/c1-9(13)3-4-11-5-7-12(8-6-11)15-10(2)14/h5-8H,3-4H2,1-2H3

InChI key

UMIKWXDGXDJQJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(3-Oxobutyl)phenyl acetate (4-(4-acetoxyphenyl) -2-butanone) is a standard melon fly attractant.[1]

Application

4-(3-Oxobutyl)phenyl acetate was used as attractant for detection programs aimed at melon fly and other cuelure-responding Bactrocera fruit flies.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric B Jang et al.
Journal of economic entomology, 100(4), 1124-1128 (2007-09-14)
Field-trapping evaluations of the new male attractant, formic acid 4-(3-oxobutyl) phenyl ester (raspberry ketone formate [RKF]) were conducted in Hawaii with wild populations of melon flies, Bactrocera cucurbitae Coquillett (Diptera: Tephritidae), to determine its activity in the field and to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service