Skip to Content
Merck
All Photos(1)

Key Documents

M26607

Sigma-Aldrich

3′-Methylacetophenone

98%

Synonym(s):

Methyl m-tolyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$103.00
25 G
$349.00

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$103.00
25 G
$349.00

About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein:
956637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

218-220 °C (lit.)

mp

−9 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccc(C)c1

InChI

1S/C9H10O/c1-7-4-3-5-9(6-7)8(2)10/h3-6H,1-2H3

InChI key

FSPSELPMWGWDRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Panić et al.
Biotechnology letters, 41(2), 253-262 (2018-12-05)
Chiral building blocks [(S)-1-(3-methylphenyl)ethanol, (S)-1-(3,4-dimethylphenyl)ethanol and (S)-1-(2,4,6-trimethylphenyl)ethanol] for drug synthesis were prepared using two green approaches: (1) the yeast Saccharomyces cerevisiae as the biocatalyst and (2) the natural deep eutectic solvents (NADES) as the alternative solvents. Three different NADES with
Huiying Song et al.
Journal of chromatography. A, 1532, 124-135 (2017-12-10)
The accuracy of the longitudinal diffusion term (b-term) plays a vital role in the study of mass transfer mechanisms in high performance liquid chromatography (HPLC). In this study, three commonly used methodologies (peak parking; fitting of an experimental van Deemter
Kunpeng Li et al.
Scientific reports, 7(1), 3081-3081 (2017-06-10)
(S)-carbonyl reductase II (SCRII) from Candida parapsilosis is a short-chain alcohol dehydrogenase/reductase. It catalyses the conversion of 2-hydroxyacetophenone to (S)-1-phenyl-1,2-ethanediol with low efficiency. Sortase was reported as a molecular "stapler" for site-specific protein conjugation to strengthen or add protein functionality.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service