Skip to Content
Merck
All Photos(1)

Key Documents

305502

Sigma-Aldrich

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAl[OC(C2H5)3]3H
CAS Number:
Molecular Weight:
380.51
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

reaction suitability

reagent type: reductant

concentration

0.5 M in THF

density

0.904 g/mL at 25 °C

SMILES string

[Li+].CCC(CC)(CC)O[AlH-](OC(CC)(CC)CC)OC(CC)(CC)CC

InChI

1S/3C7H15O.Al.Li.H/c3*1-4-7(8,5-2)6-3;;;/h3*4-6H2,1-3H3;;;/q3*-1;+2;+1;

InChI key

FXBRQZDWJRAIEB-UHFFFAOYSA-N

Application

Reactant for:
  • Stereocontrolled synthesis of sterols
  • Reduction reactions (reducing agent)
  • Stereoselective reduction of a-keto esters
  • Reduction of Weinreb amides

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service