Skip to Content
Merck
All Photos(1)

Key Documents

304123

Sigma-Aldrich

2-[(Phenylsulfonyl)methyl]benzyl bromide

97%

Synonym(s):

1-Bromomethyl-2-[(phenylsulfonyl)methyl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4CH2SO2C6H5
CAS Number:
Molecular Weight:
325.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

93-95 °C (lit.)

solubility

methanol: soluble 50 mg/mL, clear, colorless

functional group

bromo
sulfone

SMILES string

BrCc1ccccc1CS(=O)(=O)c2ccccc2

InChI

1S/C14H13BrO2S/c15-10-12-6-4-5-7-13(12)11-18(16,17)14-8-2-1-3-9-14/h1-9H,10-11H2

InChI key

SYNKYJKCQBVGSL-UHFFFAOYSA-N

Application

2-[(Phenylsulfonyl)methyl]benzyl bromide may be used in chemical synthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service