Skip to Content
Merck
All Photos(2)

Key Documents

302392

Sigma-Aldrich

Methyl 3-bromo-2-(bromomethyl)propionate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$157.00

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$157.00

About This Item

Linear Formula:
(BrCH2)2CHCO2CH3
CAS Number:
Molecular Weight:
259.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

60-62 °C/0.4 mmHg (lit.)

density

1.82 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

COC(=O)C(CBr)CBr

InChI

1S/C5H8Br2O2/c1-9-5(8)4(2-6)3-7/h4H,2-3H2,1H3

InChI key

USXVPPOARMSYGY-UHFFFAOYSA-N

Application

Methyl 3-bromo-2-(bromomethyl)propionate was used in synthesis of sultams and (S)-1-benzyl-3-hydroxypyrrolidine.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Rolfe et al.
Organic & biomolecular chemistry, 8(9), 2198-2203 (2010-04-20)
The synthesis of small organic molecules as probes for discovering new therapeutic agents has been an important aspect of chemical-biology. Herein we report a reagent-based, diversity-oriented synthetic (DOS) strategy to probe chemical and biological space via a "Click, Click, Cyclize"

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service