Skip to Content
Merck
All Photos(1)

Key Documents

299693

Sigma-Aldrich

Azelaic acid dichloride

98%

Synonym(s):

Azelaoyl chloride, Nonanedioyl chloride, Nonanedioyl dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$168.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$168.00

About This Item

Linear Formula:
ClOC(CH2)7COCl
CAS Number:
Molecular Weight:
225.11
Beilstein:
1365424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.467 (lit.)

bp

166 °C/18 mmHg (lit.)

density

1.143 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)CCCCCCCC(Cl)=O

InChI

1S/C9H14Cl2O2/c10-8(12)6-4-2-1-3-5-7-9(11)13/h1-7H2

InChI key

HGEVGSTXQGZPCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azelaic acid dichloride (Azelaoyl chloride) was used in synthesis of:
  • series of electrochromic polyamides (alternating copolymers)[1]
  • 1,9-diphenylnonane-1,9-dione[2]
  • 1,9-di(4-butylphenyl)nonane-1,9-dione[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble nickel bis (dithiolene) oligomers for third-order non-linear optical studies.
Callum AS.
Journal of Materials Chemistry, 4(8), 1233-1237 (1994)
Synthesis and Characterization of Electrochromic Polyamides with Well-Defined Molecular Structures and Redox Properties.
Gao J, et al.
Advances in Functional Materials, 14(6), 537-543 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service