Skip to Content
Merck
All Photos(3)

Key Documents

295760

Sigma-Aldrich

5-Amino-1-ethylpyrazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

92-94 °C/0.01 mmHg (lit.)

mp

52-57 °C (lit.)

SMILES string

CCn1nccc1N

InChI

1S/C5H9N3/c1-2-8-5(6)3-4-7-8/h3-4H,2,6H2,1H3

InChI key

FSJOLBAFVKSQQJ-UHFFFAOYSA-N

Application

5-Amino-1-ethylpyrazole is a heterocyclic building block[1], has been used in the preparation of 1-protected-1H-pyrazolo[3,4-b]pyridines[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrazolo [3, 4-b] pyridines. The preparation of 1-protected-1 H-pyrazolo [3, 4-b] pyridines and attempts to remove the 1-substituent. Some reactions of 1-benzyl-1 H-pyrazolo [3, 4-b] pyridine and its 7-oxide.
Dorgan RJJ, et al.
Journal of the Chemical Society. Perkin Transactions 1, 938-942 (1980)
Journal of the Chemical Society. Perkin Transactions 1, 938-938 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service