Skip to Content
Merck
All Photos(1)

Key Documents

731412

Sigma-Aldrich

6-Iodoquinoline

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$105.00

$105.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$105.00

About This Item

Empirical Formula (Hill Notation):
C9H6IN
CAS Number:
Molecular Weight:
255.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$105.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

form

solid

mp

86-90 °C

functional group

iodo

SMILES string

Ic1ccc2ncccc2c1

InChI

1S/C9H6IN/c10-8-3-4-9-7(6-8)2-1-5-11-9/h1-6H

InChI key

WKTASELJZCIVBR-UHFFFAOYSA-N

Application

Reactant for:
  • Room temperature CuI-catalyzed N-arylation using 2-pyridinyl β-ketones ligands

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service