Skip to Content
Merck
All Photos(1)

Key Documents

295124

Sigma-Aldrich

Carbonyl sulfide

≥97.5%

Synonym(s):

Carbon oxysulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
COS
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
form:
gas
Assay:
≥97.5%
Pricing and availability is not currently available.

vapor density

2.1 (20 °C, vs air)

Quality Level

vapor pressure

9034 mmHg ( 21 °C)

Assay

≥97.5%

form

gas

expl. lim.

11.9-29 %

bp

−50 °C (lit.)

mp

−138 °C (lit.)

SMILES string

O=C=S

InChI

1S/COS/c2-1-3

InChI key

JJWKPURADFRFRB-UHFFFAOYSA-N

Other Notes

For 295124-300G-EU packaging: 500 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 295124-300G-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Flam. Gas 1B - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xunchen Liu et al.
Physical chemistry chemical physics : PCCP, 13(31), 14235-14242 (2011-07-22)
We report a combined high resolution infrared and microwave spectroscopic investigation of the acetylene-ammonia and carbonyl sulfide-ammonia complexes using a pulsed slit-nozzle multipass absorption spectrometer based on a quantum cascade laser and a pulsed nozzle beam Fourier transform microwave spectrometer
Keren Stimler et al.
The New phytologist, 186(4), 869-878 (2010-03-20)
*Carbonyl sulfide (COS) exchange in C(3) leaves is linked to that of CO(2), providing a basis for the use of COS as a powerful tracer of gross CO(2) fluxes between plants and the atmosphere, a critical element in understanding the
Xian Shu Li et al.
Microbial ecology, 60(1), 96-104 (2010-06-24)
Nineteen fungal strains having an ability to oxidize elemental sulfur in mineral salts medium were isolated from deteriorated sandstones of Angkor monuments. These fungi formed clearing zone on agar medium supplemented with powder sulfur due to the dissolution of sulfur.
Takahiro Ogawa et al.
Journal of the American Chemical Society, 135(10), 3818-3825 (2013-02-15)
Carbonyl sulfide (COS) is an atmospheric trace gas leading to sulfate aerosol formation, thereby participating in the global radiation balance and ozone chemistry, but its biological sinks are not well understood. Thiobacillus thioparus strain THI115 can grow on thiocyanate (SCN(-))
J Norooz Oliaee et al.
The journal of physical chemistry. A, 114(49), 12834-12838 (2010-11-17)
Vibration-rotation spectra of the OCS-(CO(2))(2) van der Waals complex were studied by means of direct infrared absorption spectroscopy. Complexes were generated in a supersonic slit-jet apparatus, and the expansion gas was probed using a rapid-scan tunable diode laser. Infrared bands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service