Skip to Content
Merck
All Photos(1)

Key Documents

71742

Sigma-Aldrich

Sodium methanethiolate

technical, ≥90% (RT)

Synonym(s):

Methanethiol sodium salt, Sodium thiomethoxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$158.00
25 G
$589.00

$158.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$158.00
25 G
$589.00

About This Item

Linear Formula:
CH3SNa
CAS Number:
Molecular Weight:
70.09
Beilstein:
3592983
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$158.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

grade

technical

Quality Level

Assay

≥90% (RT)

form

solid

SMILES string

[Na+].C[S-]

InChI

1S/CH4S.Na/c1-2;/h2H,1H3;/q;+1/p-1

InChI key

RMBAVIFYHOYIFM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Strong nucleophile used for the synthesis of methyl aryl sulfides from halo-arenes[1][2]; Alkanethiolates are efficient reagents for SN2 dealkylation of esters and aryl ethers[3][4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Sol. 1 - Skin Corr. 1A

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Chianelli et al.
Synthesis, 475-475 (1982)
T.R. Kelly et al.
Tetrahedron Letters, 3859-3859 (1977)
L. Testaferri et al.
Synthesis, 751-751 (1983)
P. Cogolli et al.
The Journal of Organic Chemistry, 44, 2642-2642 (1979)
Nikolaos Kontoudakis et al.
Food research international (Ottawa, Ont.), 98, 95-102 (2017-06-15)
A variety of techniques have been developed with the ability to measure different forms of metals in wine with the ultimate aim of providing a more accurate indicator of metal induced spoilage of wine. This study was conducted in order

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service