Skip to Content
Merck
All Photos(3)

Key Documents

293490

Sigma-Aldrich

2-Benzylimidazoline

99%

Synonym(s):

2-Benzyl-2-imidazoline, 2-Benzyl-4,5-dihydro-1H-imidazole, Benzidazol, NSC 35110, Phenylmethylimidazoline, Tolazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

66-69 °C (lit.)

solubility

alcohol: freely soluble
chloroform: soluble
diethyl ether: very slightly soluble
ethyl acetate: very slightly soluble
water: freely soluble

functional group

phenyl

storage temp.

2-8°C

SMILES string

C1CN=C(Cc2ccccc2)N1

InChI

1S/C10H12N2/c1-2-4-9(5-3-1)8-10-11-6-7-12-10/h1-5H,6-8H2,(H,11,12)

InChI key

JIVZKJJQOZQXQB-UHFFFAOYSA-N

General description

2-Benzylimidazoline is an active vasodilator[1].

Application

2-Benzylimidazoline has been used in the preparation of phenyl-2,3-dihydroimidazo[1,2-a]pyridinone derivatives via condensation with of a series of β-keto esters[2].
Reactant for synthesis of imidazoline-containing drug

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A E Hubbell et al.
Equine veterinary journal, 38(3), 238-241 (2006-05-19)
The ability to shorten the duration of sedation would potentially improve safety and utility of detomidine. To determine the effects of tolazoline and atipamezole after detomidine sedation. Administration of tolazoline or atipamezole would not affect detomidine sedation. In a randomised
Tulio M Prado et al.
Veterinary anaesthesia and analgesia, 35(3), 201-207 (2008-02-20)
To evaluate the anesthetic and cardiorespiratory effects of two doses of intramuscular (IM) xylazine/ketamine in alpacas, and to determine if tolazoline would reduce the anesthetic recovery time. Prospective randomized crossover study. Six castrated male alpacas. Each alpaca received a low
Beulah Leitch et al.
The Journal of comparative neurology, 462(1), 55-70 (2003-05-23)
The role of the biogenic amine octopamine in modulating cholinergic synaptic transmission between the locust forewing stretch receptor neuron (fSR) and the first basalar motoneuron (BA1) was investigated. The amines 5-hydroxytryptamine (5-HT, serotonin) and dopamine were also studied. Bath application
Derivatives of phenyl-2, 3-dihydroimidazo [1, 2-a] pyridinone.
Magatti CV and Villani FJ.
Journal of Heterocyclic Chemistry, 15(6), 1021-1023 (1978)
Prevention and management of meconium aspiration syndrome--assessment of evidence based practice.
Anne Greenough et al.
European journal of pediatrics, 164(5), 329-330 (2005-02-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service