Skip to Content
Merck
All Photos(2)

Key Documents

292648

Sigma-Aldrich

Dihydrofluorescein diacetate

new

97%

Synonym(s):

Diacetyldihydrofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H18O7
CAS Number:
Molecular Weight:
418.40
Beilstein:
364167
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

mp

213-215 °C (lit.)

solubility

acetone: 50 mg/mL

fluorescence

λex 272 nm in methanol
λex 490; λem 514 (Reaction product)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(=O)Oc1ccc2C(c3ccc(OC(C)=O)cc3Oc2c1)c4ccccc4C(O)=O

InChI

1S/C24H18O7/c1-13(25)29-15-7-9-19-21(11-15)31-22-12-16(30-14(2)26)8-10-20(22)23(19)17-5-3-4-6-18(17)24(27)28/h3-12,23H,1-2H3,(H,27,28)

InChI key

YKSJJXGQHSESKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrofluorescein diacetate (H2FDA) is an esterified derivative of dihydrofluorescein. H2FDA is one of the widely used fluorescent probes employed in the intracellular reactive oxygen species (ROS) detection.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandan K. Sen, Lester Packer
Redox Cell Biology and Genetics, (2002)
Ekrem Sulukan et al.
Chemosphere, 180, 77-85 (2017-04-10)
In this study, it has been investigated that the effects of glyphosate, which is a herbicide within organophosphate and unselective widely used in agriculture on enzyme activity of carbonic anhydrase, production of reactive oxygen species, cell apoptosis and body morphology
Lehcen Benikhlef et al.
BMC plant biology, 13, 133-133 (2013-09-17)
In a previous study we have shown that wounding of Arabidopsis thaliana leaves induces a strong and transient immunity to Botrytis cinerea, the causal agent of grey mould. Reactive oxygen species (ROS) are formed within minutes after wounding and are
Hugo F Olivares-Rubio et al.
Ecotoxicology and environmental safety, 147, 840-851 (2017-10-04)
Diatoms play key roles in primary production and carbon fixation at a global scale and in some cases these species live on marine ecosystems impacted by crude oil (CO) spills. Halamphora oceanica, a new diatom species from the Southwest of
Gessica Sousa et al.
Scientific reports, 9(1), 670-670 (2019-01-27)
The termite gut is an efficient decomposer of polyphenol-rich diets, such as lignocellulosic biomasses, and it has been proposed that non-enzymatic oxidative mechanisms could be involved with the digestive process in these animals. However, oxidant levels are completely unknown in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service