Skip to Content
Merck
All Photos(1)

Documents

T5428

Sigma-Aldrich

Tetramethylrhodamine methyl ester perchlorate

≥95% purity, powder

Synonym(s):

TMRM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H25N2O3 · ClO4
CAS Number:
Molecular Weight:
500.93
Beilstein:
7162165
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Tetramethylrhodamine methyl ester perchlorate, ≥95%

Quality Level

Assay

≥95%

form

powder

composition

Carbon: 57.5-62.4%

Nitrogen: 5.3-5.9%

color

dark green

solubility

methanol: 5 mg/mL, clear

εmax

>95000 at 546-552 nm in methanol

fluorescence

λem 573 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.COC(=O)c1ccccc1C2=C3C=C\C(C=C3Oc4cc(ccc24)N(C)C)=[N+](\C)C

InChI

1S/C25H25N2O3.ClHO4/c1-26(2)16-10-12-20-22(14-16)30-23-15-17(27(3)4)11-13-21(23)24(20)18-8-6-7-9-19(18)25(28)29-5;2-1(3,4)5/h6-15H,1-5H3;(H,2,3,4,5)/q+1;/p-1

InChI key

PFYWPQMAWCYNGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylrhodamine methyl ester perchlorate (TMRM) is a membrane-permeant, cationic, fluorescent dye that can label mitochondria and mark embryonic stem cell/induced pluripotent stem cell-derived cardiomyocytes. It is useful for the isolation of mature cardiomyocytes.

Application

Tetramethylrhodamine methyl ester perchlorate (TMRM) has been used as a sensitive probe for the detection of mitochondrial membrane potential.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic Targeting of the Mitochondria Initiates Excessive Superoxide Production and Mitochondrial Depolarization Causing Decreased mtDNA Integrity
Kaytee L
PLoS ONE (2016)
Markus Hartl et al.
Molecular systems biology, 13(10), 949-949 (2017-10-25)
Histone deacetylases have central functions in regulating stress defenses and development in plants. However, the knowledge about the deacetylase functions is largely limited to histones, although these enzymes were found in diverse subcellular compartments. In this study, we determined the
Emerging Trends in Cell and Gene Therapy null
Quantitative analysis of mitochondrial morphology and membrane potential in living cells using high-content imaging, machine learning, and morphological binning
Anthony P
Biochimica et Biophysica Acta (2015)
Antigoni Diokmetzidou et al.
Journal of cell science, 129(20), 3705-3720 (2016-08-28)
The association of desmin with the α-crystallin Β-chain (αΒ-crystallin; encoded by CRYAB), and the fact that mutations in either one of them leads to heart failure in humans and mice, suggests a potential compensatory interplay between the two in cardioprotection.

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service