Skip to Content
Merck
All Photos(1)

Key Documents

289272

Sigma-Aldrich

Dysprosium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Dysprosium trichloride hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$178.00

$178.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$178.00

About This Item

Linear Formula:
DyCl3 · 6(H2O)
CAS Number:
Molecular Weight:
376.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

$178.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: dysprosium

impurities

≤2000.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.Cl[Dy](Cl)Cl

InChI

1S/3ClH.Dy.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

HFEOHRWLEGXZHW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis of a Dysprosium (III) Complex with a Hexadentate Amine Ligand: This study explores the synthesis of a novel dysprosium complex, highlighting its potential applications in fields like molecular magnetism and catalysis (Gonzalez-Barreira et al., 2023).

  • A DyIII Complex of a Pentadentate Schiff Base with Field-Induced Single-Ion Magnet Behaviour: This research describes the magnetic properties of a dysprosium complex, useful for developing new magnetic materials (Corredoira-Vazquez et al., 2023).

  • A Tetranuclear Dysprosium Schiff Base Complex Showing Slow Relaxation of Magnetization: Focuses on the synthesis and characterization of a dysprosium-based complex with potential implications in data storage technologies (Gebrezgiabher et al., 2022).

  • The self-assembly of DyF3 nanoparticles synthesized by chloride-based route: Investigates the properties of dysprosium fluoride nanoparticles synthesized using dysprosium(III) chloride hexahydrate, relevant for various nanotechnology applications (Alakshin et al., 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María N Oviedo et al.
Analytical methods : advancing methods and applications, 13(8), 1033-1042 (2021-02-09)
Efficient separation and preconcentration of inorganic Sb species in different water samples were performed in this work by a novel dispersive liquid-liquid microextraction (DLLME) method based on the application of a magnetic ionic liquid (MIL) and electrothermal atomic absorption spectroscopy

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service