Skip to Content
Merck
All Photos(1)

Key Documents

288799

Sigma-Aldrich

3′-Chloroacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCH3
CAS Number:
Molecular Weight:
154.59
Beilstein:
636318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.550 (lit.)

bp

227-229 °C (lit.)

density

1.191 g/mL at 25 °C (lit.)

functional group

chloro
ketone

SMILES string

CC(=O)c1cccc(Cl)c1

InChI

1S/C8H7ClO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,1H3

InChI key

UUWJBXKHMMQDED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The activity of strain, Saccharomyces cerevisiae B5, on the reduction of 3′-chloroacetophenone was studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhimin Ou et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 43(4), 523-526 (2005-11-10)
Four strains of microorganisms which have activity of chloroacetophenone reduction were screened, in which Saccharomyces cerevisiae B5 showed the highest activity and good stereoselectivity. This strains showed different activity on the reduction of various substrates in the following order: 2'-chloroacetophenone
Kunpeng Li et al.
Scientific reports, 7(1), 3081-3081 (2017-06-10)
(S)-carbonyl reductase II (SCRII) from Candida parapsilosis is a short-chain alcohol dehydrogenase/reductase. It catalyses the conversion of 2-hydroxyacetophenone to (S)-1-phenyl-1,2-ethanediol with low efficiency. Sortase was reported as a molecular "stapler" for site-specific protein conjugation to strengthen or add protein functionality.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service