Skip to Content
Merck
All Photos(1)

Key Documents

286680

Sigma-Aldrich

L-α-Methyltyrosine

98%

Synonym(s):

α-Methyl-L-tyrosine, L-2-Methyl-3-(4-hydroxyphenyl)alanine, L-AMPT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2C(CH3)(NH2)CO2H
CAS Number:
Molecular Weight:
195.22
Beilstein:
2368400
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]20/D −7°, c = 1 in 0.1 M HCl

SMILES string

C[C@](N)(Cc1ccc(O)cc1)C(O)=O

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjun Chen et al.
Drug discovery today, 16(15-16), 697-703 (2011-06-01)
Drug-induced liver injury (DILI) is a leading cause of drugs failing during clinical trials and being withdrawn from the market. Comparative analysis of drugs based on their DILI potential is an effective approach to discover key DILI mechanisms and risk
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service