Skip to Content
Merck
All Photos(1)

Key Documents

286656

Sigma-Aldrich

α-Methyl-DL-phenylalanine

98%

Synonym(s):

(±)-2-Amino-2-methyl-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$232.00

$232.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$232.00

About This Item

Linear Formula:
C6H5CH2C(CH3)(NH2)CO2H
CAS Number:
Molecular Weight:
179.22
Beilstein:
2803960
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$232.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

293-294 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(N)(Cc1ccccc1)C(O)=O

InChI

1S/C10H13NO2/c1-10(11,9(12)13)7-8-5-3-2-4-6-8/h2-6H,7,11H2,1H3,(H,12,13)

InChI key

HYOWVAAEQCNGLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of phenylalanine, p-chlorophenylalanine and alpha-methylphenylalanine on glucose uptake in vitro by the brain of young rats.
N R Rodrigues et al.
Biochemical Society transactions, 18(3), 419-419 (1990-06-01)
M S de Freitas et al.
Neurochemistry international, 26(4), 381-385 (1995-04-01)
We studied the effects of L-phenylalanine and alpha-methylphenylalanine on 32P in vitro incorporation into cytoskeletal proteins from cerebral cortex of 17-day-old rats. Slices of cerebral cortex were incubated in the absence or presence of increasing concentrations of L-phenylalanine, alpha-methylphenylalanine or
S Ma et al.
Journal of chromatography. A, 1216(18), 3784-3793 (2009-03-13)
Enantiomeric separation of two aromatic alpha-substituted alanine esters was achieved on two commercially available polysaccharide-based chiral stationary phases (CSPs): amylose tris(3,5-dimethylphenylcarbamate) (ADMPC) and cellulose tris(3,5-dimethylphenylcarbamate) (CDMPC). The interactions between enantiomeric analytes and the CSPs were investigated using chromatographic methods and
A Diamond et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 14(5 Pt 2), 3072-3082 (1994-05-01)
Phenylketonuria (PKU) is a genetic disorder in which the hydroxylation of phenylalanine (Phe) to tyrosine is severely disrupted. If PKU is left untreated, severe mental retardation results. The accepted treatment is to restrict dietary intake of Phe. It has generally
C A Brass et al.
The Biochemical journal, 208(3), 765-771 (1982-12-15)
Hyperphenylalaninaemia induced by daily injections of alpha-methylphenylalanine plus phenylalanine caused 20-40% decreases in cerebral dopamine (3,4-dihydroxyphenethylamine) and noradrenaline in 7- and 11-day-old rats. alpha-Methylphenylalanine alone as well as phenylalanine alone caused cerebral dopamine depletion. However, the effects were not additive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service