Skip to Content
Merck
All Photos(3)

Key Documents

284750

Sigma-Aldrich

6-Methylpyridine-3-carboxylic acid

99%

Synonym(s):

6-Methylnicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
112049
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

210-213 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1ccc(cn1)C(O)=O

InChI

1S/C7H7NO2/c1-5-2-3-6(4-8-5)7(9)10/h2-4H,1H3,(H,9,10)

InChI key

RZOKQIPOABEQAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The co-ordination complex of the formula [Ni(L1)2(H2O)4].4H2O where L1H = 6-methylpyridine-3-carboxylic acid was structurally charaterized using X-ray single crystal analysis[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3D non porous and thermally labile nickel (II) and manganese (II) complexes with hetero donor ligands: Synthesis, X-ray single crystal structure, thermal and luminescent study.
Dalai S, et al.
Inorgorganica Chimica Acta, 363(8), 1843-1848 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service